missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2,4-bis(trifluoromethyl)benzene 97.0+%, TCI America™

Catalog No. p-7123408
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B42071G 1 g
B42075G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B42071G Supplier TCI America Supplier No. B42071G

Chemical Identifiers

CAS 327-75-3
Molecular Formula C8H3BrF6
Molecular Weight (g/mol) 293.01
MDL Number MFCD00074904
InChI Key QDEJWLIKRLJYEK-UHFFFAOYSA-N
Synonym 2,4-bis trifluoromethyl bromobenzene, 1-bromo-2,4-bis trifluoromethyl benzene, 4-bromo-1,3-bis trifluoromethyl benzene, 1,3-bis trifluoromethyl-4-bromobenzene, 2,4-ditrifluoromethylbromobenzene, benzene, 1-bromo-2,4-bis trifluoromethyl, 2,4-bis-trifluoromethylbromobenzene, fxffr be exfff, pubchem8494
PubChem CID 136164
IUPAC Name 1-bromo-2,4-bis(trifluoromethyl)benzene
SMILES FC(F)(F)C1=CC=C(Br)C(=C1)C(F)(F)F

Specifications

Color Yellow
Boiling Point 51°C
Quantity 1 g
Formula Weight 293.01
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-2,4-bis(trifluoromethyl)benzene