accessibility menu, dialog, popup

UserName

CAS RN 327-75-3

IUPAC Name: 1-bromo-2,4-bis(trifluoromethyl)benzene
Synonyms: 1-bromo-2,4-bis(trifluoromethyl)benzene
Molecular Weight (g/mol): 293.01
Molecular Formula: C8H3BrF6
InChi Key: QDEJWLIKRLJYEK-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=C(Br)C(=C1)C(F)(F)F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results