missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2,4,6-trifluorobenzene 98.0+%, TCI America™

Catalog No. p-7118894
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B11685G 5 g
B116825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B11685G Supplier TCI America Supplier No. B11685G
Only null left

Chemical Identifiers

CAS 2367-76-2
Molecular Formula C6H2BrF3
Molecular Weight (g/mol) 210.98
MDL Number MFCD00000334
InChI Key PZBSPSOGEVCRQI-UHFFFAOYSA-N
Synonym 1-bromo-2,4,6-trifluorobenzene, 2,4,6-trifluorobromobenzene, 2-bromo-1,3,5-trifluoro-benzene, benzene, 2-bromo-1,3,5-trifluoro, 1-bromo-2,4,6-trifluoro benzene, 1,3,5-trifluoro-2-bromobenzene, fr cf ef be, pubchem4306, acmc-1cbzk, intermediates-zcf02629
PubChem CID 75398
IUPAC Name 2-bromo-1,3,5-trifluorobenzene
SMILES FC1=CC(F)=C(Br)C(F)=C1

Specifications

Melting Point 4°C
Color Yellow
Boiling Point 140°C
Quantity 5 g
UN Number 1993
Formula Weight 210.98
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-2,4,6-trifluorobenzene