missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromo-2,3-difluorobenzene 98.0+%, TCI America™
$54.06 - $307.57
Chemical Identifiers
| CAS | 38573-88-5 |
|---|---|
| Molecular Formula | C6H3BrF2 |
| Molecular Weight (g/mol) | 192.991 |
| MDL Number | MFCD00061136 |
| InChI Key | RKWWASUTWAFKHA-UHFFFAOYSA-N |
| Synonym | 2,3-difluorobromobenzene, 2,3-difluorobrmorobenzene, 1-bromo-2,3-difluoro-benzene, 3-bromo-1,2-difluorobenzene, difluorobromobenzene, pubchem1041, acmc-1aibi, 2,3-diflurobromobenzene, 2,3-difluorophenyl bromide, 2,3-difluoro-1-bromobenzene |
| PubChem CID | 2733260 |
| IUPAC Name | 1-bromo-2,3-difluorobenzene |
| SMILES | C1=CC(=C(C(=C1)Br)F)F |
Chemical Identifiers
| 38573-88-5 | |
| 192.991 | |
| RKWWASUTWAFKHA-UHFFFAOYSA-N | |
| 2733260 | |
| C1=CC(=C(C(=C1)Br)F)F |
| C6H3BrF2 | |
| MFCD00061136 | |
| 2,3-difluorobromobenzene, 2,3-difluorobrmorobenzene, 1-bromo-2,3-difluoro-benzene, 3-bromo-1,2-difluorobenzene, difluorobromobenzene, pubchem1041, acmc-1aibi, 2,3-diflurobromobenzene, 2,3-difluorophenyl bromide, 2,3-difluoro-1-bromobenzene | |
| 1-bromo-2,3-difluorobenzene |
Specifications
| 38573-88-5 | |
| C6H3BrF2 | |
| 1 g | |
| RKWWASUTWAFKHA-UHFFFAOYSA-N | |
| 1-bromo-2,3-difluorobenzene | |
| 2733260 | |
| ≥98.0% (GC) | |
| 1-Bromo-2,3-difluorobenzene |
| Yellow | |
| MFCD00061136 | |
| 2,3-difluorobromobenzene, 2,3-difluorobrmorobenzene, 1-bromo-2,3-difluoro-benzene, 3-bromo-1,2-difluorobenzene, difluorobromobenzene, pubchem1041, acmc-1aibi, 2,3-diflurobromobenzene, 2,3-difluorophenyl bromide, 2,3-difluoro-1-bromobenzene | |
| C1=CC(=C(C(=C1)Br)F)F | |
| 192.991 | |
| 192.99 | |
| Liquid |
Safety and Handling
TSCA : No