accessibility menu, dialog, popup

UserName

FBrF

CAS RN 38573-88-5

IUPAC Name: 1-bromo-2,3-difluorobenzene
Synonyms: 1-bromo-2,3-difluorobenzene
Molecular Weight (g/mol): 192.99
Molecular Formula: C6H3BrF2
InChi Key: RKWWASUTWAFKHA-UHFFFAOYSA-N
SMILES: FC1=CC=CC(Br)=C1F
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)