missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2,3-difluorobenzene 98.0+%, TCI America™

Catalog No. B18001G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B18001G 1 g
B18005G 5 g
B180025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B18001G Supplier TCI America Supplier No. B18001G

Chemical Identifiers

CAS 38573-88-5
Molecular Formula C6H3BrF2
Molecular Weight (g/mol) 192.991
MDL Number MFCD00061136
InChI Key RKWWASUTWAFKHA-UHFFFAOYSA-N
Synonym 2,3-difluorobromobenzene, 2,3-difluorobrmorobenzene, 1-bromo-2,3-difluoro-benzene, 3-bromo-1,2-difluorobenzene, difluorobromobenzene, pubchem1041, acmc-1aibi, 2,3-diflurobromobenzene, 2,3-difluorophenyl bromide, 2,3-difluoro-1-bromobenzene
PubChem CID 2733260
IUPAC Name 1-bromo-2,3-difluorobenzene
SMILES C1=CC(=C(C(=C1)Br)F)F

Specifications

Color Yellow
Quantity 1 g
Formula Weight 192.99
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-2,3-difluorobenzene