missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2,3-difluorobenzene, 98%

CAS: 38573-88-5 | C6H3BrF2 | 192.991 g/mol

$92.15 - $268.33

Chemical Identifiers

CAS 38573-88-5
Molecular Formula C6H3BrF2
Molecular Weight (g/mol) 192.991
MDL Number MFCD00061136
InChI Key RKWWASUTWAFKHA-UHFFFAOYSA-N
Synonym 2,3-difluorobromobenzene, 2,3-difluorobrmorobenzene, 1-bromo-2,3-difluoro-benzene, 3-bromo-1,2-difluorobenzene, difluorobromobenzene, pubchem1041, acmc-1aibi, 2,3-diflurobromobenzene, 2,3-difluorophenyl bromide, 2,3-difluoro-1-bromobenzene
PubChem CID 2733260
IUPAC Name 1-bromo-2,3-difluorobenzene
SMILES C1=CC(=C(C(=C1)Br)F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2158503
View Documents
Thermo Scientific Chemicals
B2158503
1 g
Each for $92.15
Only null left
 
AAB2158506
View Documents
Thermo Scientific Chemicals
B2158506
5 g
Each for $268.33
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

38573-88-5
192.991
RKWWASUTWAFKHA-UHFFFAOYSA-N
2733260
C1=CC(=C(C(=C1)Br)F)F
C6H3BrF2
MFCD00061136
2,3-difluorobromobenzene, 2,3-difluorobrmorobenzene, 1-bromo-2,3-difluoro-benzene, 3-bromo-1,2-difluorobenzene, difluorobromobenzene, pubchem1041, acmc-1aibi, 2,3-diflurobromobenzene, 2,3-difluorophenyl bromide, 2,3-difluoro-1-bromobenzene
1-bromo-2,3-difluorobenzene

Specifications

38573-88-5
69°C to 70°C (12 mmHg)
C6H3BrF2
MFCD00061136
2,3-difluorobromobenzene, 2,3-difluorobrmorobenzene, 1-bromo-2,3-difluoro-benzene, 3-bromo-1,2-difluorobenzene, difluorobromobenzene, pubchem1041, acmc-1aibi, 2,3-diflurobromobenzene, 2,3-difluorophenyl bromide, 2,3-difluoro-1-bromobenzene
C1=CC(=C(C(=C1)Br)F)F
192.991
192.99
1-Bromo-2,3-difluorobenzene
1.724
61°C (141°F)
1.509
1 g
RKWWASUTWAFKHA-UHFFFAOYSA-N
1-bromo-2,3-difluorobenzene
2733260
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only