missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2,3,5-trifluorobenzene 98.0+%, TCI America™

Catalog No. B29705G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B29705G 5 g
B29701G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B29705G Supplier TCI America Supplier No. B29705G
Only null left

Chemical Identifiers

CAS 133739-70-5
Molecular Formula C6H2BrF3
Molecular Weight (g/mol) 210.98
MDL Number MFCD00012232
InChI Key XSMLLZPSNLQCQU-UHFFFAOYSA-N
PubChem CID 2736399
IUPAC Name 1-bromo-2,3,5-trifluorobenzene
SMILES FC1=CC(F)=C(F)C(Br)=C1

Specifications

Color Yellow
Quantity 5 g
UN Number 1993
Synonym 2,3,5-trifluorobromobenzene, benzene, 1-bromo-2,3,5-trifluoro, 1-bromo-2,3,5-trifluoro-benzene, pubchem2305, acmc-209bte, intermediates-zcf02035, 2,3,5-trifluorobromo benzene, 2,3,5-trifluorophenyl bromide, paragos 390204, benzene,1-bromo-2,3,5-trifluoro
Formula Weight 210.98
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-2,3,5-trifluorobenzene