accessibility menu, dialog, popup

UserName

FFFBr

CAS RN 133739-70-5

IUPAC Name: 1-bromo-2,3,5-trifluorobenzene
Synonyms: 1-bromo-2,3,5-trifluorobenzene
Molecular Weight (g/mol): 210.98
Molecular Formula: C6H2BrF3
InChi Key: XSMLLZPSNLQCQU-UHFFFAOYSA-N
SMILES: FC1=CC(F)=C(F)C(Br)=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results