missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2,3,5,6-tetramethylbenzene, 99%

Catalog No. p-7043584
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0146506 5 g
AAL0146514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0146506 Supplier Thermo Scientific Chemicals Supplier No. L0146506
Only null left

CAS: 1646-53-3 | C10H13Br | 213.118 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1646-53-3
Molecular Formula C10H13Br
Molecular Weight (g/mol) 213.118
MDL Number MFCD00000070
InChI Key WJKBPTLQJXKEHC-UHFFFAOYSA-N
Synonym bromodurene, 3-bromodurene, 1-bromo-2,3,5,6-tetramethylbenzene, benzene, 3-bromo-1,2,4,5-tetramethyl, bromdurol, duryl bromide, bromodurene, pract., acmc-1bz04, 2,3,5,6-tetramethylbromobenzene, 1-bromo-2,5,6-tetramethylbenzene
PubChem CID 74240
IUPAC Name 3-bromo-1,2,4,5-tetramethylbenzene
SMILES CC1=CC(=C(C(=C1C)Br)C)C

Specifications

Melting Point 58°C to 62°C
Boiling Point 115°C to 120°C (5 mmHg)
Quantity 5 g
Beilstein 2207158
Formula Weight 213.12
Percent Purity 99%
Chemical Name or Material 1-Bromo-2,3,5,6-tetramethylbenzene
EINECSNumber 216-707-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only