missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromo-2,3,4-trifluorobenzene 98.0+%, TCI America™
$48.46 - $143.54
Chemical Identifiers
| CAS | 176317-02-5 |
|---|---|
| Molecular Formula | C6H2BrF3 |
| Molecular Weight (g/mol) | 210.981 |
| MDL Number | MFCD00013248 |
| InChI Key | MUUAQFJJUGVBGB-UHFFFAOYSA-N |
| Synonym | 2,3,4-trifluorobromobenzene, 2,3,4-trifluoro bromobenzene, benzene, 1-bromo-2,3,4-trifluoro, 1-bromo-2,3,4-trifluoro-benzene, trifluorobromobenzene, pubchem7089, trifluorophenyl bromide, fr bf cf de, acmc-209eb9, ksc497i1f |
| PubChem CID | 2733255 |
| IUPAC Name | 1-bromo-2,3,4-trifluorobenzene |
| SMILES | C1=CC(=C(C(=C1F)F)F)Br |
Chemical Identifiers
| 176317-02-5 | |
| 210.981 | |
| MUUAQFJJUGVBGB-UHFFFAOYSA-N | |
| 2733255 | |
| C1=CC(=C(C(=C1F)F)F)Br |
| C6H2BrF3 | |
| MFCD00013248 | |
| 2,3,4-trifluorobromobenzene, 2,3,4-trifluoro bromobenzene, benzene, 1-bromo-2,3,4-trifluoro, 1-bromo-2,3,4-trifluoro-benzene, trifluorobromobenzene, pubchem7089, trifluorophenyl bromide, fr bf cf de, acmc-209eb9, ksc497i1f | |
| 1-bromo-2,3,4-trifluorobenzene |
Specifications
| 176317-02-5 | |
| 60°C | |
| MFCD00013248 | |
| 2,3,4-trifluorobromobenzene, 2,3,4-trifluoro bromobenzene, benzene, 1-bromo-2,3,4-trifluoro, 1-bromo-2,3,4-trifluoro-benzene, trifluorobromobenzene, pubchem7089, trifluorophenyl bromide, fr bf cf de, acmc-209eb9, ksc497i1f | |
| C1=CC(=C(C(=C1F)F)F)Br | |
| 210.981 | |
| 210.98 | |
| Liquid |
| Yellow | |
| C6H2BrF3 | |
| 1 g | |
| MUUAQFJJUGVBGB-UHFFFAOYSA-N | |
| 1-bromo-2,3,4-trifluorobenzene | |
| 2733255 | |
| ≥98.0% (GC) | |
| 1-Bromo-2,3,4-trifluorobenzene |
Safety and Handling
TSCA : No