missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2,2-dimethoxypropane, 98%

Catalog No. AAA1113909
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1113909 10 g
AAA1113918 50 g
AAA1113930 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1113909 Supplier Thermo Scientific Chemicals Supplier No. A1113909

CAS: 126-38-5 | C5H11BrO2 | 183.045 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 126-38-5
Molecular Formula C5H11BrO2
Molecular Weight (g/mol) 183.045
MDL Number MFCD00000208
InChI Key SGTITUFGCGGICE-UHFFFAOYSA-N
Synonym propane,1-bromo-2,2-dimethoxy, propane, 1-bromo-2,2-dimethoxy, bromoacetone dimethyl acetal, bromo-2,2dimethoxypropane, bromo-2,2-dimethoxypropane, 1-bromo-2,2-dimethoxypropan, 1-bromo-2,2-dimethoxyprpopane, 1-bromo-2,2-dimethoxy propane, 1-bromo-2,2-dimethoxy-propane, bromoacetone dimethyl ketal
PubChem CID 67175
IUPAC Name 1-bromo-2,2-dimethoxypropane
SMILES CC(CBr)(OC)OC

Specifications

Density 1.354
Boiling Point 83°C to 87°C (80 mmHg)
Flash Point 40°C (104°F)
Refractive Index 1.447
Quantity 10 g
UN Number UN3271
Beilstein 2036095
Sensitivity Moisture Sensitive
Formula Weight 183.05
Percent Purity 98%
Chemical Name or Material 1-Bromo-2,2-dimethoxypropane
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHERS, N.O.S.
EINECSNumber 204-784-7
TSCA No

RUO – Research Use Only