accessibility menu, dialog, popup

UserName

CAS RN 126-38-5

IUPAC Name: 1-bromo-2,2-dimethoxypropane
Synonyms: 1-bromo-2,2-dimethoxypropane
Molecular Weight (g/mol): 183.05
Molecular Formula: C5H11BrO2
InChi Key: SGTITUFGCGGICE-UHFFFAOYSA-N
SMILES: COC(C)(CBr)OC
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)