missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-N-Fmoc-L-tryptophan, 97%

CAS: 143824-78-6 | C31H30N2O6 | 526.59 g/mol

Supplier:  Thermo Scientific Chemicals H5966314

 View more versions of this product

Catalog No. AAH5966314


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-Boc-N-Fmoc-L-tryptophan
∽97°C (decomposition)
MFCD00153366
UN3077
Moisture sensitive
ADOHASQZJSJZBT-SANMLTNESA-N
CC(C)(C)OC(=O)N1C=C(C[C@H](NC(=O)OCC2C3=C(C=CC=C3)C3=C2C=CC=C3)C(O)=O)C2=CC=CC=C12
526.59
526.58
143824-78-6
C31H30N2O6
25 g
7698009
fmoc-trp boc-oh, fmoc-l-trp boc-oh, nalpha-9h-fluoren-9-ylmethoxy carbonyl-n1-tert-butoxycarbonyl-l-tryptophan, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-1-tert-butoxycarbonyl-1h-indol-3-yl propanoic acid, 2s-3-1-tert-butoxycarbonyl indol-3-yl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, fmoc-l-trp boc, pubchem10048, 1-boc-n-fmoc-l-tryptophan, ksc910o8t, n1-boc-nalpha-fmoc-l-tryptophan
−21° (c=1 in DMF)
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[1-[(2-methylpropan-2-yl)oxycarbonyl]indol-3-yl]propanoic acid
9849766
97%

Chemical Identifiers

143824-78-6
526.59
ADOHASQZJSJZBT-SANMLTNESA-N
9849766
C31H30N2O6
MFCD00153366
fmoc-trp boc-oh, fmoc-l-trp boc-oh, nalpha-9h-fluoren-9-ylmethoxy carbonyl-n1-tert-butoxycarbonyl-l-tryptophan, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-1-tert-butoxycarbonyl-1h-indol-3-yl propanoic acid, 2s-3-1-tert-butoxycarbonyl indol-3-yl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, fmoc-l-trp boc, pubchem10048, 1-boc-n-fmoc-l-tryptophan, ksc910o8t, n1-boc-nalpha-fmoc-l-tryptophan
CC(C)(C)OC(=O)N1C=C(C[C@H](NC(=O)OCC2C3=C(C=CC=C3)C3=C2C=CC=C3)C(O)=O)C2=CC=CC=C12
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H317
May cause an allergic skin reaction.

P261-P272-P280g-P302+P352-P333+P313-P363-P501c

H317

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

TSCA : No

Recommended Storage : Keep cold; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only