missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-N-Fmoc-L-tryptophan, 97%

Catalog No. AAH5966306
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5966306 5 g
AAH5966314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5966306 Supplier Thermo Scientific Chemicals Supplier No. H5966306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 143824-78-6 | C31H30N2O6 | 526.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 143824-78-6
Molecular Formula C31H30N2O6
Molecular Weight (g/mol) 526.59
MDL Number MFCD00153366
InChI Key ADOHASQZJSJZBT-SANMLTNESA-N
Synonym fmoc-trp boc-oh, fmoc-l-trp boc-oh, nalpha-9h-fluoren-9-ylmethoxy carbonyl-n1-tert-butoxycarbonyl-l-tryptophan, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-1-tert-butoxycarbonyl-1h-indol-3-yl propanoic acid, 2s-3-1-tert-butoxycarbonyl indol-3-yl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, fmoc-l-trp boc, pubchem10048, 1-boc-n-fmoc-l-tryptophan, ksc910o8t, n1-boc-nalpha-fmoc-l-tryptophan
PubChem CID 9849766
SMILES CC(C)(C)OC(=O)N1C=C(C[C@H](NC(=O)OCC2C3=C(C=CC=C3)C3=C2C=CC=C3)C(O)=O)C2=CC=CC=C12

Specifications

Melting Point ∽97°C (decomposition)
Quantity 5 g
UN Number UN3077
Beilstein 7698009
Sensitivity Moisture sensitive
Optical Rotation −21° (c=1 in DMF)
IUPAC Name 3-{1-[(tert-butoxy)carbonyl]-1H-indol-3-yl}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid
Formula Weight 526.58
Percent Purity 97%
Chemical Name or Material 1-Boc-N-Fmoc-L-tryptophan
Hazard Category H317
Hazard Statement GHS H Statement
H317
May cause an allergic skin reaction.
Precautionary Statement P261-P272-P280g-P302+P352-P333+P313-P363-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
TSCA No
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only