missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-azetidine-3-carboxaldehyde, 97%

CAS: 177947-96-5 | C9H15NO3 | 185.223 g/mol

Supplier:  Thermo Scientific Chemicals H5279403

 View more versions of this product

Catalog No. AAH5279403


Only null left

Description

Description

1-Boc-azetidine-3-carboxaldehyde is used as a starting material in organic synthesis. It is also used as an intermediate in pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Boc-azetidine-3-carboxaldehyde is used as a starting material in organic synthesis. It is also used as an intermediate in pharmaceuticals.

Notes
Air sensitive. Store in a cool place. Incompatible with water and oxidizing agents.
Specifications

Specifications

1-Boc-azetidine-3-carboxaldehyde
77°C to 82°C (2mmHg)
MFCD06656142
JVQOZRRUGOADSU-UHFFFAOYSA-N
tert-butyl 3-formylazetidine-1-carboxylate
185.223
185.22
97%
177947-96-5
C9H15NO3
1-boc-3-azetidinecarboxaldehyde, 1-boc-3-formylazetidine, 1-boc-azetidine-3-carboxaldehyde, azetidine-3-carboxaldehyde, 1-azetidinecarboxylic acid, 3-formyl-, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl azetidine-3-carboxaldehyde, 3-formylazetidine-1-carboxylic acid tert-butylester, pubchem10136, 1-boc-3-azetidinecarbaldehyde, 1-boc-3-azetidine carboxaldehyde
CC(C)(C)OC(=O)N1CC(C1)C=O
1 g
10726182
Air sensitive

Chemical Identifiers

177947-96-5
185.223
JVQOZRRUGOADSU-UHFFFAOYSA-N
10726182
CC(C)(C)OC(=O)N1CC(C1)C=O
C9H15NO3
MFCD06656142
1-boc-3-azetidinecarboxaldehyde, 1-boc-3-formylazetidine, 1-boc-azetidine-3-carboxaldehyde, azetidine-3-carboxaldehyde, 1-azetidinecarboxylic acid, 3-formyl-, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl azetidine-3-carboxaldehyde, 3-formylazetidine-1-carboxylic acid tert-butylester, pubchem10136, 1-boc-3-azetidinecarbaldehyde, 1-boc-3-azetidine carboxaldehyde
tert-butyl 3-formylazetidine-1-carboxylate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Keep cold; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only