accessibility menu, dialog, popup

UserName

CH3H3CCH3OONO

CAS RN 177947-96-5

IUPAC Name: tert-butyl 3-formylazetidine-1-carboxylate
Synonyms: tert-butyl 3-formylazetidine-1-carboxylate
Molecular Weight (g/mol): 185.22
Molecular Formula: C9H15NO3
InChi Key: JVQOZRRUGOADSU-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CC(C1)C=O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)