missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-azetidine-3-carboxaldehyde, 97%

Catalog No. p-7035088
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52794MD 250 mg
AAH5279403 1 g
AAH5279406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH52794MD Supplier Thermo Scientific Chemicals Supplier No. H52794MD
Only null left

CAS: 177947-96-5 | C9H15NO3 | 185.223 g/mol

1-Boc-azetidine-3-carboxaldehyde is used as a starting material in organic synthesis. It is also used as an intermediate in pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Boc-azetidine-3-carboxaldehyde is used as a starting material in organic synthesis. It is also used as an intermediate in pharmaceuticals.

Notes
Air sensitive. Store in a cool place. Incompatible with water and oxidizing agents.

Chemical Identifiers

CAS 177947-96-5
Molecular Formula C9H15NO3
Molecular Weight (g/mol) 185.223
MDL Number MFCD06656142
InChI Key JVQOZRRUGOADSU-UHFFFAOYSA-N
Synonym 1-boc-3-azetidinecarboxaldehyde, 1-boc-3-formylazetidine, 1-boc-azetidine-3-carboxaldehyde, azetidine-3-carboxaldehyde, 1-azetidinecarboxylic acid, 3-formyl-, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl azetidine-3-carboxaldehyde, 3-formylazetidine-1-carboxylic acid tert-butylester, pubchem10136, 1-boc-3-azetidinecarbaldehyde, 1-boc-3-azetidine carboxaldehyde
PubChem CID 10726182
IUPAC Name tert-butyl 3-formylazetidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CC(C1)C=O

Specifications

Boiling Point 77°C to 82°C (2mmHg)
Quantity 250 mg
Formula Weight 185.22
Sensitivity Air sensitive
Percent Purity 97%
Chemical Name or Material 1-Boc-azetidine-3-carboxaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only