missing translation for 'onlineSavingsMsg'
Learn More

1-[(Benzyloxy)carbonyl]piperidine-4-carboxylic acid, 97%, Thermo Scientific™

Chemical Identifiers

CAS 10314-98-4
Molecular Formula C14H17NO4
Molecular Weight (g/mol) 263.293
MDL Number MFCD01568759
InChI Key URTPNQRAHXRPMP-UHFFFAOYSA-N
Synonym 1-benzyloxy carbonyl piperidine-4-carboxylic acid, n-cbz-4-piperidinecarboxylic acid, 1-cbz-4-piperidinecarboxylic acid, n-cbz-piperidine-4-carboxylic acid, 1-benzyloxycarbonyl piperidine-4-carboxylic acid, 1-cbz-piperidine-4-carboxylic acid, 1-benzyloxy carbonyl-4-piperidinecarboxylic acid, n-cbz-isonipecotic acid, piperidine-1,4-dicarboxylic acid monobenzyl ester
PubChem CID 736489
IUPAC Name 1-phenylmethoxycarbonylpiperidine-4-carboxylic acid
SMILES C1CN(CCC1C(=O)O)C(=O)OCC2=CC=CC=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
CC07501DA
View Documents Promotion Details
N/A 1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC07501FA
View Documents Promotion Details
N/A 100g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

10314-98-4
263.293
URTPNQRAHXRPMP-UHFFFAOYSA-N
736489
C1CN(CCC1C(=O)O)C(=O)OCC2=CC=CC=C2
C14H17NO4
MFCD01568759
1-benzyloxy carbonyl piperidine-4-carboxylic acid, n-cbz-4-piperidinecarboxylic acid, 1-cbz-4-piperidinecarboxylic acid, n-cbz-piperidine-4-carboxylic acid, 1-benzyloxycarbonyl piperidine-4-carboxylic acid, 1-cbz-piperidine-4-carboxylic acid, 1-benzyloxy carbonyl-4-piperidinecarboxylic acid, n-cbz-isonipecotic acid, piperidine-1,4-dicarboxylic acid monobenzyl ester
1-phenylmethoxycarbonylpiperidine-4-carboxylic acid

Specifications

10314-98-4
100.0
C14H17NO4
1-benzyloxy carbonyl piperidine-4-carboxylic acid, n-cbz-4-piperidinecarboxylic acid, 1-cbz-4-piperidinecarboxylic acid, n-cbz-piperidine-4-carboxylic acid, 1-benzyloxycarbonyl piperidine-4-carboxylic acid, 1-cbz-piperidine-4-carboxylic acid, 1-benzyloxy carbonyl-4-piperidinecarboxylic acid, n-cbz-isonipecotic acid, piperidine-1,4-dicarboxylic acid monobenzyl ester
C1CN(CCC1C(=O)O)C(=O)OCC2=CC=CC=C2
263.293
263.29
1-[(Benzyloxy)carbonyl]piperidine-4-carboxylic acid
96.5
97%
MFCD01568759
URTPNQRAHXRPMP-UHFFFAOYSA-N
1-phenylmethoxycarbonylpiperidine-4-carboxylic acid
736489
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents