missing translation for 'onlineSavingsMsg'
Learn More

1-[(Benzyloxy)carbonyl]piperidine-4-carboxylic acid, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC07501FA

Quantity 100g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC07501FA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

10314-98-4
263.293
URTPNQRAHXRPMP-UHFFFAOYSA-N
736489
C1CN(CCC1C(=O)O)C(=O)OCC2=CC=CC=C2
C14H17NO4
MFCD01568759
1-benzyloxy carbonyl piperidine-4-carboxylic acid, n-cbz-4-piperidinecarboxylic acid, 1-cbz-4-piperidinecarboxylic acid, n-cbz-piperidine-4-carboxylic acid, 1-benzyloxycarbonyl piperidine-4-carboxylic acid, 1-cbz-piperidine-4-carboxylic acid, 1-benzyloxy carbonyl-4-piperidinecarboxylic acid, n-cbz-isonipecotic acid, piperidine-1,4-dicarboxylic acid monobenzyl ester
1-phenylmethoxycarbonylpiperidine-4-carboxylic acid

Specifications

1-[(Benzyloxy)carbonyl]piperidine-4-carboxylic acid
96.5
97%
C14H17NO4
MFCD01568759
URTPNQRAHXRPMP-UHFFFAOYSA-N
1-phenylmethoxycarbonylpiperidine-4-carboxylic acid
736489
97%
10314-98-4
100.0
Amber Glass Bottle
100g
1-benzyloxy carbonyl piperidine-4-carboxylic acid, n-cbz-4-piperidinecarboxylic acid, 1-cbz-4-piperidinecarboxylic acid, n-cbz-piperidine-4-carboxylic acid, 1-benzyloxycarbonyl piperidine-4-carboxylic acid, 1-cbz-piperidine-4-carboxylic acid, 1-benzyloxy carbonyl-4-piperidinecarboxylic acid, n-cbz-isonipecotic acid, piperidine-1,4-dicarboxylic acid monobenzyl ester
C1CN(CCC1C(=O)O)C(=O)OCC2=CC=CC=C2
263.293
263.29
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents