accessibility menu, dialog, popup

UserName

OONOO

CAS RN 10314-98-4

IUPAC Name: 1-[(benzyloxy)carbonyl]piperidine-4-carboxylate
Synonyms: 1-[(benzyloxy)carbonyl]piperidine-4-carboxylate
Molecular Weight (g/mol): 262.29
Molecular Formula: C14H16NO4
InChi Key: URTPNQRAHXRPMP-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1CCN(CC1)C(=O)OCC1=CC=CC=C1
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)