missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyloxy-4-iodobenzene 98.0+%, TCI America™

Catalog No. p-7123692
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B46925G 5 g
B469225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B46925G Supplier TCI America Supplier No. B46925G
Only null left

Chemical Identifiers

CAS 19578-68-8
Molecular Formula C13H11IO
Molecular Weight (g/mol) 310.134
MDL Number MFCD00079704
InChI Key MPWFGAWFTAZWKZ-UHFFFAOYSA-N
PubChem CID 519671
IUPAC Name 1-iodo-4-phenylmethoxybenzene
SMILES C1=CC=C(C=C1)COC2=CC=C(C=C2)I

Specifications

Melting Point 62°C
Color White
Boiling Point 140°C
Quantity 5 g
Synonym 1-benzyloxy-4-iodobenzene, 4-benzyloxyiodobenzene, 4-iodobenzyloxybenzene, benzyl 4-iodophenyl ether, benzene, 1-iodo-4-phenylmethoxy, 1-benzyloxy-4-iodo-benzene, benzene,1-iodo-4-phenylmethoxy, acmc-20amon, 4-benzyloxy iodobenzene
Formula Weight 310.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Benzyloxy-4-iodobenzene