missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Benzyloxy-4-iodobenzene, 98+%
Description
1-Benzyloxy-4-iodobenzene is used as an emulgator and as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Benzyloxy-4-iodobenzene is used as an emulgator and as a pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to light. Incompatible with oxidizing agents.
1-Benzyloxy-4-iodobenzene is used as an emulgator and as a pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to light. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | 60°C to 64°C |
| Boiling Point | 140°C (1 mmHg) |
| Quantity | 5 g |
| Beilstein | 3051495 |
| Sensitivity | Light sensitive |
| Synonym | 1-benzyloxy-4-iodobenzene, 4-benzyloxyiodobenzene, 4-iodobenzyloxybenzene, benzyl 4-iodophenyl ether, benzene, 1-iodo-4-phenylmethoxy, 1-benzyloxy-4-iodo-benzene, benzene,1-iodo-4-phenylmethoxy, acmc-20amon, 4-benzyloxy iodobenzene |
| Solubility Information | Insoluble in water. |
| Formula Weight | 310.14 |
| Percent Purity | ≥98% |
| Chemical Name or Material | 1-Benzyloxy-4-iodobenzene |
Safety and Handling
| EINECSNumber | 000-000-0 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only