missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyloxy-3-iodobenzene, 99%

CAS: 107623-21-2 | C13H11IO | 310.13 g/mol

Supplier:  Thermo Scientific Chemicals L1744203

 View more versions of this product

Catalog No. AAL1744203


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-Benzyloxy-3-iodobenzene
50°C to 51°C
MFCD01318100
3270112
1-benzyloxy-3-iodobenzene, 3-iodobenzyloxybenzene, 1-benzyloxy-3-iodo-benzene, benzene,1-iodo-3-phenylmethoxy, benzene, 1-iodo-3-phenylmethoxy, pubchem3066, 3-benzyloxyiodobenzene, acmc-1c7zz, 1-benzyl-oxy-3-iodobenzene
IC1=CC=CC(OCC2=CC=CC=C2)=C1
310.13
310.14
107623-21-2
C13H11IO
1 g
Light sensitive
QMKHOPJXDQAHBG-UHFFFAOYSA-N
1-iodo-3-phenylmethoxybenzene
4228019
99%

Chemical Identifiers

107623-21-2
310.13
QMKHOPJXDQAHBG-UHFFFAOYSA-N
4228019
IC1=CC=CC(OCC2=CC=CC=C2)=C1
C13H11IO
MFCD01318100
1-benzyloxy-3-iodobenzene, 3-iodobenzyloxybenzene, 1-benzyloxy-3-iodo-benzene, benzene,1-iodo-3-phenylmethoxy, benzene, 1-iodo-3-phenylmethoxy, pubchem3066, 3-benzyloxyiodobenzene, acmc-1c7zz, 1-benzyl-oxy-3-iodobenzene
1-iodo-3-phenylmethoxybenzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only