missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyl-3-pyrroline, 98%

CAS: 6913-92-4 | C11H13N | 159.232 g/mol

Supplier:  Thermo Scientific Chemicals L1949606

 View more versions of this product

Catalog No. AAL1949606


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-Benzyl-3-pyrroline
0.837
92°C (197°F)
1.542
5 g
111101
1-benzyl-3-pyrroline, 1-benzyl-2,5-dihydro-1h-pyrrole, n-benzylpyrroline, n-benzyl-3-pyrroline, 1h-pyrrole, 2,5-dihydro-1-phenylmethyl, maleimide-related compound 16, l-benzyl-3-pyrroline, n-benzyl 3-pyrroline, 1-benzylpyrroline
C1C=CCN1CC2=CC=CC=C2
159.232
159.23
6913-92-4
60°C (1 mmHg)
C11H13N
MFCD00012324
UN2735
Air sensitive
LRFHKHHUKGZIGE-UHFFFAOYSA-N
1-benzyl-2,5-dihydropyrrole
561506
≥98%

Chemical Identifiers

6913-92-4
159.232
LRFHKHHUKGZIGE-UHFFFAOYSA-N
561506
C1C=CCN1CC2=CC=CC=C2
C11H13N
MFCD00012324
1-benzyl-3-pyrroline, 1-benzyl-2,5-dihydro-1h-pyrrole, n-benzylpyrroline, n-benzyl-3-pyrroline, 1h-pyrrole, 2,5-dihydro-1-phenylmethyl, maleimide-related compound 16, l-benzyl-3-pyrroline, n-benzyl 3-pyrroline, 1-benzylpyrroline
1-benzyl-2,5-dihydropyrrole
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H227-H314-H335

DOTInformation : Hazard Class: 8; Packaging Group: II

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Keep cold; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only