missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyl-3-pyrroline, 98%

Catalog No. p-7047599
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1949603 1 g
AAL1949606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1949603 Supplier Thermo Scientific Chemicals Supplier No. L1949603

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6913-92-4 | C11H13N | 159.232 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6913-92-4
Molecular Formula C11H13N
Molecular Weight (g/mol) 159.232
MDL Number MFCD00012324
InChI Key LRFHKHHUKGZIGE-UHFFFAOYSA-N
Synonym 1-benzyl-3-pyrroline, 1-benzyl-2,5-dihydro-1h-pyrrole, n-benzylpyrroline, n-benzyl-3-pyrroline, 1h-pyrrole, 2,5-dihydro-1-phenylmethyl, maleimide-related compound 16, l-benzyl-3-pyrroline, n-benzyl 3-pyrroline, 1-benzylpyrroline
PubChem CID 561506
IUPAC Name 1-benzyl-2,5-dihydropyrrole
SMILES C1C=CCN1CC2=CC=CC=C2

Specifications

Density 0.837
Boiling Point 60°C (1 mmHg)
Flash Point 92°C (197°F)
Refractive Index 1.542
Quantity 1 g
UN Number UN2735
Beilstein 111101
Sensitivity Air sensitive
Formula Weight 159.23
Percent Purity ≥98%
Chemical Name or Material 1-Benzyl-3-pyrroline
Show More Show Less
Hazard Category H227-H314-H335
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 000-000-0
TSCA No
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only