missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyl-3-pyrrolidone 98.0+%, TCI America™

Catalog No. p-7123040
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B360625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B360625G Supplier TCI America Supplier No. B360625G

Chemical Identifiers

CAS 775-16-6
Molecular Formula C11H13NO
Molecular Weight (g/mol) 175.231
MDL Number MFCD00005342
InChI Key DHGMDHQNUNRMIN-UHFFFAOYSA-N
Synonym 1-benzyl-3-pyrrolidinone, n-benzyl-3-pyrrolidinone, 1-phenylmethyl-3-pyrrolidinone, 1-benzyl-3-pyrrolidone, 1-benzyl-pyrrolidin-3-one, 1-benzyl pyrrolidin-3-one, 3-pyrrolidinone, 1-phenylmethyl, benzyl-3-pyrrolidinone, pubchem10007, n-benzylpyrrolid-3-one
PubChem CID 69890
IUPAC Name 1-benzylpyrrolidin-3-one
SMILES C1CN(CC1=O)CC2=CC=CC=C2

Specifications

Boiling Point 109°C
Quantity 25 g
Formula Weight 175.23
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-Benzyl-3-pyrrolidone
TSCA No
Recommended Storage Refrigerator