CAS RN 775-16-6
CAS RN 775-16-6
IUPAC Name:
1-benzyl-3-oxopyrrolidin-1-ium
Synonyms:
1-benzyl-3-oxopyrrolidin-1-ium
Molecular Weight (g/mol):
176.24
Molecular Formula:
C11H14NO
InChi Key:
DHGMDHQNUNRMIN-UHFFFAOYNA-O
SMILES:
O=C1CC[NH+](CC2=CC=CC=C2)C1
1
–
1
of
1
results
1
1-Benzyl-3-pyrrolidone 98.0+%, TCI America™
CAS: 775-16-6 Molecular Formula: C11H13NO Molecular Weight (g/mol): 175.231 MDL Number: MFCD00005342 InChI Key: DHGMDHQNUNRMIN-UHFFFAOYSA-N Synonym: 1-benzyl-3-pyrrolidinone,n-benzyl-3-pyrrolidinone,1-phenylmethyl-3-pyrrolidinone,1-benzyl-3-pyrrolidone,1-benzyl-pyrrolidin-3-one,1-benzyl pyrrolidin-3-one,3-pyrrolidinone, 1-phenylmethyl,benzyl-3-pyrrolidinone,pubchem10007,n-benzylpyrrolid-3-one PubChem CID: 69890 IUPAC Name: 1-benzylpyrrolidin-3-one SMILES: C1CN(CC1=O)CC2=CC=CC=C2