Learn More
1-Benzyl-1,4,7,10-tetraazacyclododecane
CAS: 112193-83-6 | C15H26N4 | 262.401 g/mol
$337.18 - $827.93
Chemical Identifiers
| CAS | 112193-83-6 |
|---|---|
| Molecular Formula | C15H26N4 |
| Molecular Weight (g/mol) | 262.401 |
| MDL Number | MFCD09263310 |
| InChI Key | FURLCQRFFWBENR-UHFFFAOYSA-N |
| Synonym | 1-benzyl-1,4,7,10-tetraazacyclododecane, 1-benzylcyclen, acmc-20aoy3, 1,4,7,10-tetraazacyclododecane, 1-phenylmethyl, n-benzyl-1,4,7,10-tetraazacyclododecane,min |
| PubChem CID | 10084082 |
| IUPAC Name | 1-benzyl-1,4,7,10-tetrazacyclododecane |
| SMILES | C1CNCCN(CCNCCN1)CC2=CC=CC=C2 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH26525MD
|
Thermo Scientific Chemicals
H26525MD |
250 mg |
Each for $337.18
|
|
|||||
|
AAH2652503
|
Thermo Scientific Chemicals
H2652503 |
1 g |
Each for $827.93
|
|
|||||
Description
Used as ligands for catalysts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsUsed as ligands for catalysts.
Solubility
Soluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Store under dry inert gas. It is hygroscopic in nature. Incompatible with oxidizing agents.
Chemical Identifiers
| 112193-83-6 | |
| 262.401 | |
| FURLCQRFFWBENR-UHFFFAOYSA-N | |
| 10084082 | |
| C1CNCCN(CCNCCN1)CC2=CC=CC=C2 |
| C15H26N4 | |
| MFCD09263310 | |
| 1-benzyl-1,4,7,10-tetraazacyclododecane, 1-benzylcyclen, acmc-20aoy3, 1,4,7,10-tetraazacyclododecane, 1-phenylmethyl, n-benzyl-1,4,7,10-tetraazacyclododecane,min | |
| 1-benzyl-1,4,7,10-tetrazacyclododecane |
Specifications
| 112193-83-6 | |
| C15H26N4 | |
| 250 mg | |
| 1-benzyl-1,4,7,10-tetraazacyclododecane, 1-benzylcyclen, acmc-20aoy3, 1,4,7,10-tetraazacyclododecane, 1-phenylmethyl, n-benzyl-1,4,7,10-tetraazacyclododecane,min | |
| FURLCQRFFWBENR-UHFFFAOYSA-N | |
| 1-benzyl-1,4,7,10-tetrazacyclododecane | |
| 10084082 | |
| 1-Benzyl-1,4,7,10-tetraazacyclododecane |
| 84°C to 86°C | |
| MFCD09263310 | |
| Hygroscopic | |
| Soluble in water. | |
| C1CNCCN(CCNCCN1)CC2=CC=CC=C2 | |
| 262.401 | |
| 262.4 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only