Learn More
1-Benzyl-1,4,7,10-tetraazacyclododecane
CAS: 112193-83-6 | C15H26N4 | 262.401 g/mol
Supplier: Thermo Scientific Chemicals H2652503
Description
Used as ligands for catalysts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsUsed as ligands for catalysts.
Solubility
Soluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Store under dry inert gas. It is hygroscopic in nature. Incompatible with oxidizing agents.
Specifications
| 1-Benzyl-1,4,7,10-tetraazacyclododecane | |
| 84°C to 86°C | |
| MFCD09263310 | |
| Hygroscopic | |
| Soluble in water. | |
| C1CNCCN(CCNCCN1)CC2=CC=CC=C2 | |
| 262.401 | |
| 262.4 |
| 112193-83-6 | |
| C15H26N4 | |
| 1 g | |
| 1-benzyl-1,4,7,10-tetraazacyclododecane, 1-benzylcyclen, acmc-20aoy3, 1,4,7,10-tetraazacyclododecane, 1-phenylmethyl, n-benzyl-1,4,7,10-tetraazacyclododecane,min | |
| FURLCQRFFWBENR-UHFFFAOYSA-N | |
| 1-benzyl-1,4,7,10-tetrazacyclododecane | |
| 10084082 |
Chemical Identifiers
| 112193-83-6 | |
| 262.401 | |
| FURLCQRFFWBENR-UHFFFAOYSA-N | |
| 10084082 | |
| C1CNCCN(CCNCCN1)CC2=CC=CC=C2 |
| C15H26N4 | |
| MFCD09263310 | |
| 1-benzyl-1,4,7,10-tetraazacyclododecane, 1-benzylcyclen, acmc-20aoy3, 1,4,7,10-tetraazacyclododecane, 1-phenylmethyl, n-benzyl-1,4,7,10-tetraazacyclododecane,min | |
| 1-benzyl-1,4,7,10-tetrazacyclododecane |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only