missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Benzhydrylpiperazine 98.0+%, TCI America™
Supplier: TCI America B376025G
Specifications
| 1-Benzhydrylpiperazine | |
| 93°C | |
| 185°C | |
| MFCD00038379 | |
| 1-diphenylmethyl piperazine, benzhydrylpiperazine, n-benzhydrylpiperazine, 1-benzhydryl-piperazine, diphenylmethylpiperazine, norcyclizine, normethylcyclizine, piperazine, 1-diphenylmethyl, 4-benzhydrylpiperazine, n-diphenylmethyl piperazine | |
| C1CN(CCN1)C(C2=CC=CC=C2)C3=CC=CC=C3 | |
| 252.361 | |
| 252.36 | |
| Crystalline Powder |
| 841-77-0 | |
| White | |
| C17H20N2 | |
| 25 g | |
| NWVNXDKZIQLBNM-UHFFFAOYSA-N | |
| 1-benzhydrylpiperazine | |
| 70048 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 841-77-0 | |
| 252.361 | |
| NWVNXDKZIQLBNM-UHFFFAOYSA-N | |
| 70048 | |
| C1CN(CCN1)C(C2=CC=CC=C2)C3=CC=CC=C3 |
| C17H20N2 | |
| MFCD00038379 | |
| 1-diphenylmethyl piperazine, benzhydrylpiperazine, n-benzhydrylpiperazine, 1-benzhydryl-piperazine, diphenylmethylpiperazine, norcyclizine, normethylcyclizine, piperazine, 1-diphenylmethyl, 4-benzhydrylpiperazine, n-diphenylmethyl piperazine | |
| 1-benzhydrylpiperazine |
Safety and Handling
RTECSNumber : TL6465000
TSCA : No