missing translation for 'onlineSavingsMsg'
Learn More

1-Benzhydrylpiperazine 98.0+%, TCI America™

Catalog No. p-7123151
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B37605G 5 g
B376025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B37605G Supplier TCI America Supplier No. B37605G
Only null left

Chemical Identifiers

CAS 841-77-0
Molecular Formula C17H20N2
Molecular Weight (g/mol) 252.361
MDL Number MFCD00038379
InChI Key NWVNXDKZIQLBNM-UHFFFAOYSA-N
Synonym 1-diphenylmethyl piperazine, benzhydrylpiperazine, n-benzhydrylpiperazine, 1-benzhydryl-piperazine, diphenylmethylpiperazine, norcyclizine, normethylcyclizine, piperazine, 1-diphenylmethyl, 4-benzhydrylpiperazine, n-diphenylmethyl piperazine
PubChem CID 70048
IUPAC Name 1-benzhydrylpiperazine
SMILES C1CN(CCN1)C(C2=CC=CC=C2)C3=CC=CC=C3

Specifications

Melting Point 93°C
Color White
Boiling Point 185°C
Quantity 5 g
Formula Weight 252.36
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Benzhydrylpiperazine
RTECSNumber TL6465000
TSCA No