missing translation for 'onlineSavingsMsg'
Learn More

1-Benzhydryl-3-azetidinol, 97%

Catalog No. AAH6672022
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6672022 100 g
AAH6672014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6672022 Supplier Thermo Scientific Chemicals Supplier No. H6672022

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 18621-17-5 | C16H17NO | 239.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 18621-17-5
Molecular Formula C16H17NO
Molecular Weight (g/mol) 239.32
MDL Number MFCD00205109
InChI Key MMAJXKGUZYDTHV-UHFFFAOYSA-N
Synonym 1-benzhydrylazetan-3-ol, 1-diphenylmethyl-3-hydroxyazetidine, 1-diphenylmethyl azetidin-3-ol, 1-benzhydryl-3-azetidinol, 1-benzhydryl-3-hydroxyazetidine, 1-benzhydryl-azetidin-3-ol, n-benzhydrylazetidin-3-ol, 3-azetidinol, 1-diphenylmethyl, 1-diphenylmethyl-3-azetidinol
PubChem CID 330448
IUPAC Name 1-(diphenylmethyl)azetidin-3-ol
SMILES OC1CN(C1)C(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Quantity 100 g
Beilstein 1534826
Formula Weight 239.32
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-Benzhydryl-3-azetidinol
Hazard Category H314-H335
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only