accessibility menu, dialog, popup

UserName

CAS RN 18621-17-5

IUPAC Name: 1-(diphenylmethyl)azetidin-3-ol
Synonyms: 1-(diphenylmethyl)azetidin-3-ol
Molecular Weight (g/mol): 239.32
Molecular Formula: C16H17NO
InChi Key: MMAJXKGUZYDTHV-UHFFFAOYSA-N
SMILES: OC1CN(C1)C(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)