missing translation for 'onlineSavingsMsg'
Learn More

1-(Azidomethyl)pyrene 98.0+%, TCI America™

Catalog No. p-7118077
Change view
Click to view available options
Quantity:
1 g
200 mg
Catalog No. Quantity
A2791200MG 200 mg
A27911G 1 g
2 options

Catalog No. A2791200MG

Supplier: TCI America A2791200MG

Only null left

Chemical Identifiers

CAS 1006061-57-9
Molecular Formula C17H11N3
Molecular Weight (g/mol) 257.296
InChI Key WLKXPQFZTSRGHP-UHFFFAOYSA-N
PubChem CID 89183461
IUPAC Name diazonio(pyren-1-ylmethyl)azanide
SMILES C1=CC2=C3C(=C1)C=CC4=C(C=CC(=C43)C=C2)C[N-][N+]#N

Specifications

Melting Point 68°C
Color Yellow
Quantity 200 mg
Formula Weight 257.30
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1-(Azidomethyl)pyrene
TSCA No
Recommended Storage Refrigerator