accessibility menu, dialog, popup

UserName

NNN

CAS RN 1006061-57-9

IUPAC Name: (diazyn-1-ium-1-yl)[(pyren-1-yl)methyl]azanide
Synonyms: diazyn-1-ium-1-yl(pyren-1-ylmethyl)azanide
Molecular Weight (g/mol): 257.3
Molecular Formula: C17H11N3
InChi Key: WLKXPQFZTSRGHP-UHFFFAOYSA-N
SMILES: N#[N+][N-]CC1=C2C=CC3=CC=CC4=CC=C(C=C1)C2=C34
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)