missing translation for 'onlineSavingsMsg'
Learn More

1-Azaphenothiazine 98.0+%, TCI America™

Catalog No. A2862200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A2862200MG 200 mg
A28621G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A2862200MG Supplier TCI America Supplier No. A2862200MG

Chemical Identifiers

CAS 261-96-1
Molecular Formula C11H8N2S
Molecular Weight (g/mol) 200.259
MDL Number MFCD00160449
InChI Key UKDZROJJLPDLDO-UHFFFAOYSA-N
Synonym 10H-Pyrido[3,2-b][1,4]benzothiazine
PubChem CID 67496
IUPAC Name 10H-pyrido[3,2-b][1,4]benzothiazine
SMILES C1=CC=C2C(=C1)NC3=C(S2)C=CC=N3

Specifications

Melting Point 112°C
Color Yellow
Boiling Point 370°C
Quantity 200 mg
Formula Weight 200.26
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Azaphenothiazine