accessibility menu, dialog, popup

UserName

CAS RN 261-96-1

HNSN

CAS RN 261-96-1

IUPAC Name: 9-thia-2,4-diazatricyclo[8.4.0.0³,⁸]tetradeca-1(14),3,5,7,10,12-hexaene
Synonyms: 9-thia-2,4-diazatricyclo[8.4.0.0³,⁸]tetradeca-1(14),3,5,7,10,12-hexaene
Molecular Weight (g/mol): 200.26
Molecular Formula: C11H8N2S
InChi Key: UKDZROJJLPDLDO-UHFFFAOYSA-N
SMILES: N1C2=CC=CC=C2SC2=CC=CN=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results