missing translation for 'onlineSavingsMsg'
Learn More

1-Aza-18-crown-6, 95%

CAS: 33941-15-0 | C12H25NO5 | 263.334 g/mol

$156.95 - $1638.21

Chemical Identifiers

CAS 33941-15-0
Molecular Formula C12H25NO5
Molecular Weight (g/mol) 263.334
MDL Number MFCD00075466
InChI Key NBXKUSNBCPPKRA-UHFFFAOYSA-N
PubChem CID 118578
IUPAC Name 1,4,7,10,13-pentaoxa-16-azacyclooctadecane
SMILES C1COCCOCCOCCOCCOCCN1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH51065MD
View Documents
Thermo Scientific Chemicals
H51065MD
250 mg
Each for $156.95
Only null left
 
AAH5106503
View Documents
Thermo Scientific Chemicals
H5106503
1 g
Each for $453.68
Only null left
 
AAH5106506
View Documents
Thermo Scientific Chemicals
H5106506
5 g
Each for $1,638.21
Only null left
 
Description

Description

1-Aza-18-crown-6 is used as building block for the synthesis of bis-crown ethers, lariat ethers and other monofunctionalized macrocycles. It is also used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Aza-18-crown-6 is used as building block for the synthesis of bis-crown ethers, lariat ethers and other monofunctionalized macrocycles. It is also used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Air sensitive. Store away from oxidizing agents.
Specifications

Chemical Identifiers

33941-15-0
263.334
NBXKUSNBCPPKRA-UHFFFAOYSA-N
1,4,7,10,13-pentaoxa-16-azacyclooctadecane
C12H25NO5
MFCD00075466
118578
C1COCCOCCOCCOCCOCCN1

Specifications

33941-15-0
C12H25NO5
250 mg
Air Sensitive
Insoluble in water.
C1COCCOCCOCCOCCOCCN1
263.334
263.33
1-Aza-18-crown-6
46°C to 49°C
MFCD00075466
1074079
1-aza-18-crown-6, 1-aza-18-crown6-ether, aza-18-crown-6, acmc-209tw0, 1-aza-18-crown 6-ether, 1-aza-18-crown nt, 1-aza-4,7,10,13,16-pentaoxacyclooctadecane
NBXKUSNBCPPKRA-UHFFFAOYSA-N
1,4,7,10,13-pentaoxa-16-azacyclooctadecane
118578
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H332-H315-H319-H335

EINECSNumber : 251-751-8

TSCA : No

Recommended Storage : Ambient temperatures; Store under Argon

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only