Learn More
1-Aza-18-crown-6, 95%
CAS: 33941-15-0 | C12H25NO5 | 263.334 g/mol
Supplier: Thermo Scientific Chemicals H5106503
Description
1-Aza-18-crown-6 is used as building block for the synthesis of bis-crown ethers, lariat ethers and other monofunctionalized macrocycles. It is also used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Aza-18-crown-6 is used as building block for the synthesis of bis-crown ethers, lariat ethers and other monofunctionalized macrocycles. It is also used as pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Air sensitive. Store away from oxidizing agents.
Specifications
| 1-Aza-18-crown-6 | |
| 46°C to 49°C | |
| MFCD00075466 | |
| 1074079 | |
| 1-aza-18-crown-6, 1-aza-18-crown6-ether, aza-18-crown-6, acmc-209tw0, 1-aza-18-crown 6-ether, 1-aza-18-crown nt, 1-aza-4,7,10,13,16-pentaoxacyclooctadecane | |
| NBXKUSNBCPPKRA-UHFFFAOYSA-N | |
| 1,4,7,10,13-pentaoxa-16-azacyclooctadecane | |
| 118578 | |
| 95% |
| 33941-15-0 | |
| C12H25NO5 | |
| 1 g | |
| Air Sensitive | |
| Insoluble in water. | |
| C1COCCOCCOCCOCCOCCN1 | |
| 263.334 | |
| 263.33 |
Chemical Identifiers
| 33941-15-0 | |
| 263.334 | |
| NBXKUSNBCPPKRA-UHFFFAOYSA-N | |
| 1,4,7,10,13-pentaoxa-16-azacyclooctadecane |
| C12H25NO5 | |
| MFCD00075466 | |
| 118578 | |
| C1COCCOCCOCCOCCOCCN1 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H332-H315-H319-H335
EINECSNumber : 251-751-8
TSCA : No
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only