missing translation for 'onlineSavingsMsg'
Learn More

1-Aza-18-crown-6, 95%

CAS: 33941-15-0 | C12H25NO5 | 263.334 g/mol

Supplier:  Thermo Scientific Chemicals H5106506

 View more versions of this product

Catalog No. AAH5106506


Only null left
Explore available promotions

Description

Description

1-Aza-18-crown-6 is used as building block for the synthesis of bis-crown ethers, lariat ethers and other monofunctionalized macrocycles. It is also used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Aza-18-crown-6 is used as building block for the synthesis of bis-crown ethers, lariat ethers and other monofunctionalized macrocycles. It is also used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Air sensitive. Store away from oxidizing agents.
Specifications

Specifications

1-Aza-18-crown-6
46°C to 49°C
MFCD00075466
1074079
1-aza-18-crown-6, 1-aza-18-crown6-ether, aza-18-crown-6, acmc-209tw0, 1-aza-18-crown 6-ether, 1-aza-18-crown nt, 1-aza-4,7,10,13,16-pentaoxacyclooctadecane
NBXKUSNBCPPKRA-UHFFFAOYSA-N
1,4,7,10,13-pentaoxa-16-azacyclooctadecane
118578
95%
33941-15-0
C12H25NO5
5 g
Air Sensitive
Insoluble in water.
C1COCCOCCOCCOCCOCCN1
263.334
263.33

Chemical Identifiers

33941-15-0
263.334
NBXKUSNBCPPKRA-UHFFFAOYSA-N
1,4,7,10,13-pentaoxa-16-azacyclooctadecane
C12H25NO5
MFCD00075466
118578
C1COCCOCCOCCOCCOCCN1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H332-H315-H319-H335

EINECSNumber : 251-751-8

TSCA : No

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only