missing translation for 'onlineSavingsMsg'
Learn More

1-Aminopyrene 98.0+%, TCI America™

Catalog No. p-7116977
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A06321G 1 g
A06325G 5 g
A063225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A06321G Supplier TCI America Supplier No. A06321G
Only null left

Chemical Identifiers

CAS 1606-67-3
Molecular Formula C16H11N
Molecular Weight (g/mol) 217.27
MDL Number MFCD00004140
InChI Key YZVWKHVRBDQPMQ-UHFFFAOYSA-N
Synonym 1-aminopyrene, 1-pyrenamine, 3-aminopyrene, pyrene, amino, pyren-1-ylamine, aminopyrene, pyrenamine, alpha-aminopyrene, unii-luw9eo1681, ccris 760
PubChem CID 15352
IUPAC Name pyren-1-amine
SMILES NC1=CC=C2C=CC3=CC=CC4=CC=C1C2=C34

Specifications

Melting Point 118°C
Quantity 1 g
Formula Weight 217.27
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Aminopyrene
EINECSNumber (4)-0389
RTECSNumber UR2275000
TSCA Yes