missing translation for 'onlineSavingsMsg'
Learn More

1-Amino-8-naphthol-2,4-disulfonic Acid Monosodium Salt Hydrate 85.0+%, TCI America™

Catalog No. p-7116857
Change view
Click to view available options
Quantity:
25 g
500 g
Catalog No. Quantity
A036325G 25 g
A0363500G 500 g
2 options

Catalog No. A036325G

Supplier: TCI America A036325G

Only null left

Chemical Identifiers

CAS 52789-62-5
Molecular Formula C10H7NNa2O7S2
Molecular Weight (g/mol) 363.27
MDL Number MFCD11042663
InChI Key XZGHMYIWMXYELB-UHFFFAOYSA-L
Synonym 4-Amino-5-hydroxy-1,3-naphthalenedisulfonic Acid Monosodium Salt, Chicago Acid, 2S Acid, Sodium Hydrogen 4-Amino-5-hydroxy-1,3-naphthalenedisulfonate, SS Acid, Monosodium 1-Amino-8-naphthol-2,4-disulfonate
PubChem CID 101310392
IUPAC Name disodium 4-amino-5-hydroxynaphthalene-1,3-disulfonate
SMILES [Na+].[Na+].NC1=C2C(O)=CC=CC2=C(C=C1S([O-])(=O)=O)S([O-])(=O)=O

Specifications

Color Black-Gray
Quantity 25 g
Formula Weight 341.28
Percent Purity ≥85.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Amino-8-naphthol-2,4-disulfonic Acid Monosodium Salt Hydrate
EINECSNumber (4)-0524
TSCA No