accessibility menu, dialog, popup

UserName

CAS RN 52789-62-5

IUPAC Name: disodium 4-amino-5-hydroxynaphthalene-1,3-disulfonate
Synonyms: disodium 4-amino-5-hydroxynaphthalene-1,3-disulfonate
Molecular Weight (g/mol): 363.27
Molecular Formula: C10H7NNa2O7S2
InChi Key: XZGHMYIWMXYELB-UHFFFAOYSA-L
SMILES: [Na+].[Na+].NC1=C2C(O)=CC=CC2=C(C=C1S([O-])(=O)=O)S([O-])(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results