missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Amino-2-butanol 98.0+%, TCI America™
$134.21 - $531.25
Chemical Identifiers
| CAS | 13552-21-1 |
|---|---|
| Molecular Formula | C4H11NO |
| Molecular Weight (g/mol) | 89.138 |
| MDL Number | MFCD00059196 |
| InChI Key | KODLUXHSIZOKTG-UHFFFAOYSA-N |
| PubChem CID | 26107 |
| IUPAC Name | 1-aminobutan-2-ol |
| SMILES | CCC(CN)O |
Chemical Identifiers
| 13552-21-1 | |
| 89.138 | |
| KODLUXHSIZOKTG-UHFFFAOYSA-N | |
| 1-aminobutan-2-ol |
| C4H11NO | |
| MFCD00059196 | |
| 26107 | |
| CCC(CN)O |
Specifications
| 13552-21-1 | |
| 169°C | |
| MFCD00059196 | |
| 2735 | |
| CCC(CN)O | |
| 89.138 | |
| 89.14 | |
| Liquid |
| Colorless | |
| C4H11NO | |
| 5 mL | |
| KODLUXHSIZOKTG-UHFFFAOYSA-N | |
| 1-aminobutan-2-ol | |
| 26107 | |
| ≥98.0% (GC,T) | |
| 1-Amino-2-butanol |
Safety and Handling
TSCA : Yes