accessibility menu, dialog, popup

UserName

CAS RN 13552-21-1

CH3HOH2N

CAS RN 13552-21-1

IUPAC Name: 1-aminobutan-2-ol
Synonyms: 1-amino-2-butanol
Molecular Weight (g/mol): 89.14
Molecular Formula: C4H11NO
InChi Key: KODLUXHSIZOKTG-UHFFFAOYNA-N
SMILES: CCC(O)CN
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results