missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Amino-2-butanol 98.0+%, TCI America™
Supplier: TCI America A082325ML
Specifications
| 1-Amino-2-butanol | |
| Colorless | |
| C4H11NO | |
| 25 mL | |
| KODLUXHSIZOKTG-UHFFFAOYSA-N | |
| 1-aminobutan-2-ol | |
| 26107 | |
| ≥98.0% (GC,T) |
| 13552-21-1 | |
| 169°C | |
| MFCD00059196 | |
| 2735 | |
| CCC(CN)O | |
| 89.138 | |
| 89.14 | |
| Liquid |
Chemical Identifiers
| 13552-21-1 | |
| 89.138 | |
| KODLUXHSIZOKTG-UHFFFAOYSA-N | |
| 1-aminobutan-2-ol |
| C4H11NO | |
| MFCD00059196 | |
| 26107 | |
| CCC(CN)O |
Safety and Handling
TSCA : Yes