missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Allyl N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-aspartate 98.0+%, TCI America™
Supplier: TCI America A28941G
Specifications
| 1-Allyl N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-aspartate | |
| 133°C | |
| C22H21NO6 | |
| 1 g | |
| ZJMVIWUCCRKNHY-IBGZPJMESA-N | |
| (3S)-3-(9H-fluoren-9-ylmethoxycarbonylamino)-4-oxo-4-prop-2-enoxybutanoic acid | |
| 7020604 | |
| ≥98.0% (HPLC,T) |
| 144120-53-6 | |
| White | |
| MFCD00467715 | |
| fmoc-asp-oall, fmoc-l-aspartic acid 1-allyl ester, s-3-9h-fluoren-9-yl methoxy carbonyl amino-4-allyloxy-4-oxobutanoic acid, 3s-3-9h-fluoren-9-ylmethoxy carbonyl amino-4-oxo-4-prop-2-en-1-yloxy butanoic acid, fmocasp oh oallyl, ambotzfaa1355, fmoc-asp-allyl oh, fmoc-asp oh-oallyl, 1-allyl n-fmoc-l-aspartate, fmoc-l-aspartic acid a-allyl ester | |
| C=CCOC(=O)C(CC(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13 | |
| 395.411 | |
| 395.41 | |
| Crystalline Powder |
Chemical Identifiers
| 144120-53-6 | |
| 395.411 | |
| ZJMVIWUCCRKNHY-IBGZPJMESA-N | |
| 7020604 | |
| C=CCOC(=O)C(CC(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13 |
| C22H21NO6 | |
| MFCD00467715 | |
| fmoc-asp-oall, fmoc-l-aspartic acid 1-allyl ester, s-3-9h-fluoren-9-yl methoxy carbonyl amino-4-allyloxy-4-oxobutanoic acid, 3s-3-9h-fluoren-9-ylmethoxy carbonyl amino-4-oxo-4-prop-2-en-1-yloxy butanoic acid, fmocasp oh oallyl, ambotzfaa1355, fmoc-asp-allyl oh, fmoc-asp oh-oallyl, 1-allyl n-fmoc-l-aspartate, fmoc-l-aspartic acid a-allyl ester | |
| (3S)-3-(9H-fluoren-9-ylmethoxycarbonylamino)-4-oxo-4-prop-2-enoxybutanoic acid |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator