missing translation for 'onlineSavingsMsg'
Learn More

1-Allyl N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-aspartate 98.0+%, TCI America™

Catalog No. p-7118132
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A28941G 1 g
A28945G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A28941G Supplier TCI America Supplier No. A28941G
Only null left

Chemical Identifiers

CAS 144120-53-6
Molecular Formula C22H21NO6
Molecular Weight (g/mol) 395.411
MDL Number MFCD00467715
InChI Key ZJMVIWUCCRKNHY-IBGZPJMESA-N
Synonym fmoc-asp-oall, fmoc-l-aspartic acid 1-allyl ester, s-3-9h-fluoren-9-yl methoxy carbonyl amino-4-allyloxy-4-oxobutanoic acid, 3s-3-9h-fluoren-9-ylmethoxy carbonyl amino-4-oxo-4-prop-2-en-1-yloxy butanoic acid, fmocasp oh oallyl, ambotzfaa1355, fmoc-asp-allyl oh, fmoc-asp oh-oallyl, 1-allyl n-fmoc-l-aspartate, fmoc-l-aspartic acid a-allyl ester
PubChem CID 7020604
IUPAC Name (3S)-3-(9H-fluoren-9-ylmethoxycarbonylamino)-4-oxo-4-prop-2-enoxybutanoic acid
SMILES C=CCOC(=O)C(CC(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13

Specifications

Melting Point 133°C
Color White
Quantity 1 g
Formula Weight 395.41
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Allyl N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-aspartate
TSCA No
Recommended Storage Refrigerator