accessibility menu, dialog, popup

UserName

H2COOOOHNHOO

CAS RN 144120-53-6

IUPAC Name: 3-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-oxo-4-(prop-2-en-1-yloxy)butanoic acid
Synonyms: 3-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-4-oxo-4-(prop-2-en-1-yloxy)butanoic acid
Molecular Weight (g/mol): 395.41
Molecular Formula: C22H21NO6
InChi Key: ZJMVIWUCCRKNHY-UHFFFAOYNA-N
SMILES: OC(=O)CC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(=O)OCC=C
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)